missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 18 Rabbit, anti-Human, Clone: 4F2, Proteintech

Product Code. p-200050435 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18048115 100 μL 100µL
18038205 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18048115 Supplier Proteintech Supplier No. 828175RR100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

The Cytokeratin 18 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Cytokeratin 18. This antibody recognizes human, mouse, rat antigen. The Cytokeratin 18 antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Antigen Cytokeratin 18
Applications Western Blot, Immunohistochemistry, ELISA
Clone 4F2
Concentration 500 μg/mL
Gene Symbols CK 18, CYK18, Cytokeratin 18, K18, keratin 18, KRT18
Host Species Rabbit
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 3875
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.