missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 19 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-687B
This item is not returnable.
View return policy
Description
Cytokeratin 19 Polyclonal specifically detects Cytokeratin 19 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Cytokeratin 19 | |
| Polyclonal | |
| Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| P08727 | |
| KRT19 | |
| Synthetic peptide derived from the C-terminal region of human Cytokeratin 19 (between residues 350-400) [UniProt P08727] | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Neuroscience, Stem Cell Markers | |
| 3880 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| CK19, CK-19, cytokeratin 19, Cytokeratin-19,40-kDa keratin intermediate filament, K19cytokeratin-19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, keratin-19, MGC15366 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse, and monkey. Rat reactivity reported in scientific literature (PMID: 25401473). | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction