missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 19 Antibody (KRT19/800), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47946H
This item is not returnable.
View return policy
Description
Cytokeratin 19 Monoclonal antibody specifically detects Cytokeratin 19 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Intracellular), Immunofluorescence, CyTOF
Specifications
| Cytokeratin 19 | |
| Monoclonal | |
| HRP | |
| 40-kDa keratin intermediate filament, CK19, CK-19, cytokeratin 19, K19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, Keratin-19, MGC15366 | |
| Recombinant full-length human Cytokeratin 19 protein. (Uniprot: P08727 ) | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Neuroscience, Stem Cell Markers | |
| 3880 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| KRT19/800 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction