missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 19 Antibody (RCK108) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-97712
This item is not returnable.
View return policy
Description
Cytokeratin 19 Monoclonal specifically detects Cytokeratin 19 in Human, Rat, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications
| Cytokeratin 19 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:1000, Flow Cytometry 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen 1:100-1:200, CyTOF-ready | |
| KRT19 | |
| RCK108 is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a cytoskeletal preparation of human bladder carcinoma cell line T24. | |
| 0.1 mg | |
| Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Neuroscience, Stem Cell Markers | |
| 3880 | |
| Human, Rat, Zebrafish | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| RCK108 | |
| Unconjugated | |
| CK19, CK-19, cytokeratin 19, Cytokeratin-19,40-kDa keratin intermediate filament, K19cytokeratin-19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, keratin-19, MGC15366 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The monoclonal antibody specifically reacts with the cytokeratin 19, a 40 kDa polypeptide, which is found in most simple and non-keratinized stratified epithelia. This antibody reacts on formalin fixed, paraffin embedded material and can be used in diagnostic immunopathology. The reaction pattern of cytokeratin 19 is documented in the literature. Antigen origin: human bladder cell line T24. Antigen location: Cytoplasm. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction