missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 19 Antibody (rKRT19/9108) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-24053JF635
This item is not returnable.
View return policy
Description
Cytokeratin 19 Monoclonal antibody specifically detects Cytokeratin 19 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin 19 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| 40-kDa keratin intermediate filament, CK19, CK-19, cytokeratin 19, K19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, Keratin-19, MGC15366 | |
| Recombinant full-length human Cytokeratin 19 protein. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Neuroscience, Stem Cell Markers | |
| 3880 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| rKRT19/9108 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction