missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 19 Mouse, Clone: A53-B/A2.26 + BA17, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3880-MSM3-P1
This item is not returnable.
View return policy
Specifications
| Cytokeratin 19 | |
| Cytoplasmic | |
| A53-B/A2.26 + BA17 | |
| Unconjugated | |
| NP_002267 | |
| Mouse | |
| 40kDa | |
| Protein A/G Purified | |
| RUO | |
| 3880 | |
| IgG2a κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| KRT19 | |
| Human breast cancer MCF-7 cells (A53-B/A2); Human mammary epithelial organoids (BA17) | |
| MCF-7, HeLa, Hep-G2 cells, Breast Cancer | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction