missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 19 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119073
This item is not returnable.
View return policy
Description
Cytokeratin 19 Polyclonal Antibody for Western Blot, IHC (P)
Cytokeratin 19 is part of a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight from 40-68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. For example, cytokeratin 19 is not expressed in hepatocytes, therefore, it is useful in the identification of liver metastasis.
Specifications
| Cytokeratin 19 | |
| Polyclonal | |
| Unconjugated | |
| KRT19 | |
| 40-kDa keratin intermediate filament; AI663979; C K1CS; CK 19; CK19; CK-19; cytokeratin 19; cytokeratin-19; EndoC; K19; K1CS; Ka19; keratin 19; keratin 19, type I; keratin complex 1, acidic, gene 19; keratin, type I cytoskeletal 19; keratin, type I, 40-kd; keratin-19; Krt-1.19; Krt1-19; Krt19; LOC100344434; MGC15366; Type I keratin Ka19 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100515339, 360626, 3880 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P08727, Q63279 | |
| KRT19 | |
| Recombinant protein Cytokeratin 19. The antigen corresponds to amino acid range 245-391 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Rat, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction