missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 20 Antibody (10D8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42616
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Cytokeratin 20 Monoclonal specifically detects Cytokeratin 20 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Cytokeratin 20 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:3000, Flow Cytometry 1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:200 | |
| NP_061883 | |
| KRT20 | |
| Recombinant human KRT20 (1-424aa) purified from E.coli. | |
| 100 μL | |
| Cytoskeleton Markers | |
| 54474 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 10D8 | |
| Unconjugated | |
| PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| CD20, CK20, CK-20, cytokeratin-20, K20cytokeratin 20, keratin 20, keratin, type I cytoskeletal 20, keratin-20, KRT21, MGC35423, Protein IT | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction