missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 20 Antibody (17A11), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26177-50ul
This item is not returnable.
View return policy
Description
Cytokeratin 20 Monoclonal antibody specifically detects Cytokeratin 20 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry
Specifications
| Cytokeratin 20 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| 17A11 | |
| Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| CD20, CK20, CK-20, cytokeratin-20, K20cytokeratin 20, keratin 20, keratin, type I cytoskeletal 20, keratin-20, KRT21, MGC35423, Protein IT | |
| A synthesized peptide derived from Human Cytokeratin 20 [UniProt P35900] | |
| 50 μL | |
| Cell Biology, Cellular Markers | |
| 54474 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction