missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582875
This item is not returnable.
View return policy
Description
Immunogen sequence: MDFSRRSFHR SLSSSLQAPV VSTVGMQRLG TTPSVYGGAG GRGIRISNSR HTVNYGSDLT GGGDLFVGNE KMAMQNLND Highest antigen sequence indentity to the following orthologs: Mouse - 63%, Rat - 69%.
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. This cytokeratin is a major cellular protein of mature enterocytes and goblet cells and is specifically expressed in the gastric and intestinal mucosa. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.
Specifications
| Cytokeratin 20 | |
| Polyclonal | |
| Unconjugated | |
| KRT20 | |
| 9030623C06Rik; CD20; CK20; CK-20; CK-21; cytokeratin 20; cytokeratin 21; cytokeratin-20; Cytokeratin-21; K20; keratin 20; keratin 20, type I; keratin, type I cytoskeletal 20; Keratin-20; krt20; Krt21; MGC35423; Protein IT | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54474 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P35900 | |
| KRT20 | |
| Recombinant protein corresponding to Human KRT20. Recombinant protein control fragment (Product #RP-93445). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction