Learn More
Invitrogen™ Cytokeratin 20 Recombinant Rabbit Monoclonal Antibody (17A11)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560695
Product Code. 30534641
  • € 459.00 / 100µL
Description
Cytokeratin 20 Recombinant Monoclonal Antibody for ELISA, Flow, IHC (P)
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. This cytokeratin is a major cellular protein of mature enterocytes and goblet cells and is specifically expressed in the gastric and intestinal mucosa. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cytokeratin 20
Recombinant Monoclonal
0.2 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P35900
KRT20
A synthesized peptide derived from Human KRT20.
100 μL
Primary
Human
Antibody
IgG
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin)
17A11
Unconjugated
KRT20
9030623C06Rik; CD20; CK20; CK-20; CK-21; cytokeratin 20; cytokeratin 21; cytokeratin-20; Cytokeratin-21; K20; keratin 20; keratin 20, type I; keratin, type I cytoskeletal 20; Keratin-20; krt20; Krt21; MGC35423; Protein IT
Rabbit
Affinity chromatography
RUO
54474
-20°C or -80°C if preferred