missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 3 Antibody (KRT3/2542), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08582JF549
This item is not returnable.
View return policy
Description
Cytokeratin 3 Monoclonal antibody specifically detects Cytokeratin 3 in Human, Bovine, Rabbit samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin 3 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| CK3, CK-3, cytokeratin-3, FLJ95909, K3cytokeratin 3, keratin 3, keratin, type II cytoskeletal 3,65 kDa cytokeratin, Keratin-3, Type-II keratin Kb3 | |
| Tissue, cells or virus corresponding to Rabbit Cytokeratin 3/CK-3. Rabbit corneal epithelial keratin. | |
| 100 μg | |
| Cell Biology, Cytoskeleton Markers, Immunology, Neuroscience, Signal Transduction | |
| 3850 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| KRT3/2542 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Bovine, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction