missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 5/6 Antibody (KRT5.6/2438), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08622CL1
This item is not returnable.
View return policy
Description
Cytokeratin 5/6 Monoclonal antibody specifically detects Cytokeratin 5/6 in Human samples. It is validated for Immunohistochemistry (Paraffin),Protein Array
Specifications
| Cytokeratin 5/6 | |
| Monoclonal | |
| CoraFluor 1 | |
| 58 kDa cytokeratin, CK5, CK-5, cytokeratin-5, DDD, EBS2, epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types, K5, keratin 5, keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockaynetypes), keratin, type II cytoskeletal 5, Keratin-5, KRT5A, Type-II keratin Kb5 | |
| Recombinant full-length human Cytokeratin 5/6 protein | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 3852 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Protein Array | |
| KRT5.6/2438 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction