missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 5/6 Antibody (KRT5.6/8275R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 311.00 - € 553.00
Specifications
| Antigen | Cytokeratin 5/6 |
|---|---|
| Clone | KRT5.6/8275R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18629743
|
Novus Biologicals
NBP3-20538-100ug |
100 μg |
€ 553.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654864
|
Novus Biologicals
NBP3-20538-20ug |
20 μg |
€ 311.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 5/6 Monoclonal antibody specifically detects Cytokeratin 5/6 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Cytokeratin 5/6 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers | |
| 10mM PBS with 0.05% BSA | |
| 3852 | |
| IgG κ | |
| Protein A or G purified |
| KRT5.6/8275R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 58 kDa cytokeratin, CK5, CK-5, cytokeratin-5, DDD, EBS2, epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types, K5, keratin 5, keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockaynetypes), keratin, type II cytoskeletal 5, Keratin-5, KRT5A, Type-II keratin Kb5 | |
| Recombinant fragments of human Cytokeratin 5 (KRT5) and Cytokeratin 6 proteins (exact sequences are proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title