missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 5/8 Antibody (SPM268), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47823CL1
This item is not returnable.
View return policy
Description
Cytokeratin 5/8 Monoclonal antibody specifically detects Cytokeratin 5/8 in Bovine,Canine,Chicken (Negative),Ferret,Hamster,Human,Mouse,Porcine,Rat,Sheep,Xenopus (Negative) samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunofluorescence,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| Cytokeratin 5/8 | |
| Monoclonal | |
| CoraFluor 1 | |
| cell proliferation-inducing protein 46, CK18, CK5, CYK18, DDD, K18, K1CO, K5, Keratin, Keratin 18, Keratin 5, KRT18, krt5, KRT5A, KRTB, type I cytoskeletal 18, type II cytoskeletal 5 | |
| Cytoskeletal preparation from HeLa cells | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 3852 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
| SPM268 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Bovine, Canine, Chicken (Negative), Ferret, Hamster | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction