missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 5 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95276F
This item is not returnable.
View return policy
Description
Cytokeratin 5 Polyclonal specifically detects Cytokeratin 5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| Cytokeratin 5 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| 58 kDa cytokeratin, CK5, CK-5, cytokeratin-5, DDD, EBS2, epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types, K5, keratin 5, keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockaynetypes), keratin, type II cytoskeletal 5, Keratin-5, KRT5A, Type-II keratin Kb5 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Cytokeratin 5. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 3852 | |
| Human | |
| Purified |
| Immunofluorescence | |
| FITC | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human Cytokeratin 5/KRT5 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction