Learn More
Cytokeratin 5 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-28506-100UL
Product Code. 17462533
  • € 450.00 / 100µL
Description
The Cytokeratin 5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Cytokeratin 5. This antibody recognizes human, mouse, rat antigen. The Cytokeratin 5 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifications
Specifications
Cytokeratin 5
Polyclonal
Rabbit
Primary
Human, Rat, Mouse
Immunofluorescence, Flow Cytometry (Intracellular Staining)
CoraLite 488
100 μL
3852