Learn More
Cytokeratin 5 Rabbit anti-Human, CoraLite 555, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL555-28506-100UL
Product Code. 17531646
  • € 450.00 / 100µL
Description
The Cytokeratin 5 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human Cytokeratin 5. This antibody recognizes human, mouse, rat antigen. The Cytokeratin 5 antibody has been validated for the following applications: IF analysis. This antibody is coralite™555.
Specifications
Specifications
Cytokeratin 5
Polyclonal
Rabbit
Primary
Human, Mouse, Rat
Immunofluorescence
CoraLite 555
100 μL
3852