missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 6 Antibody (KRT6/8267R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-23895-100ug
This item is not returnable.
View return policy
Description
Cytokeratin 6 Monoclonal antibody specifically detects Cytokeratin 6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin 6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Allergen Hom s 5, CK6A, CK-6A, CK6C, CK6D, CK-6D, cytokeratin 6A, cytokeratin 6C, cytokeratin 6D, Cytokeratin-6A, Cytokeratin-6D, K6A, K6C, K6D, keratin 6A, keratin 6C, keratin 6D, keratin, epidermal type II, K6A, keratin, type II cytoskeletal 6A, Keratin-6A, KRT6C, KRT6D, Type-II keratin Kb6 | |
| Recombinant fragment (around aa1-200) of human Cytokeratin 6 (exact sequence is proprietary) | |
| 100 μg | |
| Cell Biology, Cellular Markers, Stem Cell Markers | |
| 3853 | |
| Store at 4°C. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| KRT6/8267R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction