missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 6 Antibody (KRT6B/2116), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08612AF750
This item is not returnable.
View return policy
Description
Cytokeratin 6 Monoclonal antibody specifically detects Cytokeratin 6 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin 6 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Allergen Hom s 5, CK6A, CK-6A, CK6C, CK6D, CK-6D, cytokeratin 6A, cytokeratin 6C, cytokeratin 6D, Cytokeratin-6A, Cytokeratin-6D, K6A, K6C, K6D, keratin 6A, keratin 6C, keratin 6D, keratin, epidermal type II, K6A, keratin, type II cytoskeletal 6A, Keratin-6A, KRT6C, KRT6D, Type-II keratin Kb6 | |
| Recombinant full-length human Cytokeratin 6 protein (Uniprot: P04259 ) | |
| 100 μg | |
| Cell Biology, Cellular Markers, Stem Cell Markers | |
| 3853 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Immunohistochemistry (Paraffin) | |
| KRT6B/2116 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction