missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 6 Antibody (LHK6 (same as LHK6B)), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34669FR
This item is not returnable.
View return policy
Description
Cytokeratin 6 Monoclonal specifically detects Cytokeratin 6 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Cytokeratin 6 | |
| Monoclonal | |
| DyLight 680 | |
| Allergen Hom s 5, CK6A, CK-6A, CK6C, CK6D, CK-6D, cytokeratin 6A, cytokeratin 6C, cytokeratin 6D, Cytokeratin-6A, Cytokeratin-6D, K6A, K6C, K6D, keratin 6A, keratin 6C, keratin 6D, keratin, epidermal type II, K6A, keratin, type II cytoskeletal 6A, Keratin-6A, KRT6C, KRT6D, Type-II keratin Kb6 | |
| Mouse | |
| 56 kDa | |
| 0.1 mL | |
| Cytoskeleton Markers, Stem Cell Markers | |
| 3853 | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| LHK6 (same as LHK6B) | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| KRT6A | |
| A synthetic peptide of 11 amino acids (GSSTIKYTTTS) from C-terminus of human keratin 6 | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This monoclonal antibody recognizes a protein of 56kDa, identified as cytokeratin 6 (CK6). In humans, multiple isoforms of Cytokeratin 6 (6A-6F), encoded by several highly homologous genes, have distinct tissue expression patterns, and Cytokeratin 6A is the dominant form in epithelial tissue. The gene encoding human Cytokeratin 6A maps to chromosome 12q13, and mutations in this gene are linked to several inheritable hair and skin pathologies. Keratins 6 and 16 are expressed in keratinocytes, which are undergoing rapid turnover in the suprabasal region (also known as hyper-proliferation-related keratins). Keratin 6 is found in hair follicles, suprabasal cells of a variety of internal stratified epithelia, in epidermis, in both normal and hyper-proliferative situations. Epidermal injury results in activation of keratinocytes, which express CK6 and CK16. CK6 is strongly expressed in about 75% of head and neck squamous cell carcinomas. Expression of CK6 is particularly associated with differentiation. | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction