Learn More
Cytokeratin 6 Antibody (SPM269), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-34358-0.1mg
 View more versions of this product
Product Code. 18460602
  • € 509.00 / 0.10mg
Description
Cytokeratin 6 Monoclonal antibody specifically detects Cytokeratin 6 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cytokeratin 6
Monoclonal
0.2 mg/mL
Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL
P02538, P04259, P48668
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse
Purified
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
SPM269
Unconjugated
10 mM PBS with 0.05% BSA
Allergen Hom s 5, CK6A, CK-6A, CK6C, CK6D, CK-6D, cytokeratin 6A, cytokeratin 6C, cytokeratin 6D, Cytokeratin-6A, Cytokeratin-6D, K6A, K6C, K6D, keratin 6A, keratin 6C, keratin 6D, keratin, epidermal type II, K6A, keratin, type II cytoskeletal 6A, Keratin-6A, KRT6C, KRT6D, Type-II keratin Kb6
A synthetic peptide of 11 amino acids (GSSTIKYTTTS) from C-terminus of human Cytokeratin 6
0.1 mg
Cell Biology, Cellular Markers, Stem Cell Markers
3853
Store at 4C.
IgG2a κ