missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 6a Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 369.00 - € 560.70
Specifications
| Antigen | Cytokeratin 6a |
|---|---|
| Dilution | Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625866
|
Novus Biologicals
NBP2-49325-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18638456
|
Novus Biologicals
NBP2-49325 |
0.1 mL |
€ 593.00 € 560.70 / 0.10mL Save € 32.30 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 6a Polyclonal antibody specifically detects Cytokeratin 6a in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Cytokeratin 6a | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers, Stem Cell Markers | |
| PBS (pH 7.2), 40% Glycerol | |
| 3853 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Allergen Hom s 5, CK6A, CK-6A, CK6C, CK6D, CK-6D, cytokeratin 6A, cytokeratin 6C, cytokeratin 6D, Cytokeratin-6A, Cytokeratin-6D, K6A, K6C, K6D, keratin 6A, keratin 6C, keratin 6D, keratin, epidermal type II, K6A, keratin, type II cytoskeletal 6A, Keratin-6A, KRT6C, KRT6D, Type-II keratin Kb6 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: SGRAIGGGLSSVGGGSSTIKYTTTSSSSRKSYKH | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title