Learn More
Cytokeratin 6a Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31443
This item is not returnable.
View return policy
Description
Cytokeratin 6a Polyclonal specifically detects Cytokeratin 6a in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Immunohistochemistry | |
| Recombinant protein encompassing a sequence within the center region of human Cytokeratin 6. The exact sequence is proprietary. | |
| Affinity Purified |
| Rabbit | |
| 60 kDa | |
| RUO |