Learn More
Cytokeratin 6a Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-31443
Product Code. 18755633
  • € 555.00 / 0.10mL
Description
Cytokeratin 6a Polyclonal specifically detects Cytokeratin 6a in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunohistochemistry
Recombinant protein encompassing a sequence within the center region of human Cytokeratin 6. The exact sequence is proprietary.
Affinity Purified
Rabbit
60 kDa