Learn More
Cytokeratin 6A Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-10590-100UL
Product Code. 17492463
  • € 450.00 / 100µL
Description
The Cytokeratin 6A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Cytokeratin 6A. This antibody recognizes human, mouse, rat antigen. The Cytokeratin 6A antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™488.
Specifications
Specifications
Cytokeratin 6A
Polyclonal
Rabbit
Primary
Human, Rat, Mouse
Flow Cytometry (Intracellular Staining)
CoraLite 488
100 μL
3853