missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 6C Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
€ 180.00 - € 453.00
Specifications
| Antigen | Cytokeratin 6C |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17753444
|
Invitrogen™
PA5122047 |
20 μL |
€ 180.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17743444
![]() |
Invitrogen™
PA5122046 |
100 μL |
€ 453.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KRT6C (Keratin 6C) is a Protein Coding gene. Diseases associated with KRT6C include Palmoplantar Keratoderma, Nonepidermolytic, Focal Or Diffuse and Focal Palmoplantar Keratoderma. Among its related pathways are Cytoskeletal Signaling and Cytoskeleton remodeling Neurofilaments. Gene Ontology (GO) annotations related to this gene include structural molecule activity. An important paralog of this gene is KRT6B.Specifications
| Cytokeratin 6C | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P48668 | |
| 286887 | |
| Glu163-Gly482 (Accession P48668), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| KRT6C |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Cytokeratin 6C; KRT6C | |
| KRT6C | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title