missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 7/13/17/18 Antibody (LdS 23), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2776H
This item is not returnable.
View return policy
Description
Cytokeratin 7/13/17/18 Monoclonal specifically detects Cytokeratin 7/13/17/18 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Cytokeratin 7/13/17/18 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| KRT7 | |
| Cytoskeletons of the human urothelial RT4 cell line | |
| 0.1 mL | |
| Cytoskeleton Markers | |
| 3855 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| LdS 23 | |
| Western Blot, Immunohistochemistry | |
| CK7, CK-7, cytokeratin 7, Cytokeratin-7, K2C7, K7keratin, 55K type II cytoskeletal, keratin 7, keratin, type II cytoskeletal 7, keratin-7, MGC129731, MGC3625, Sarcolectin, SCLkeratin, simple epithelial type I, K7, type II mesothelial keratin K7, Type-II keratin Kb7 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction