missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 7/17 Antibody (C-46), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08343JF549
This item is not returnable.
View return policy
Description
Cytokeratin 7/17 Monoclonal antibody specifically detects Cytokeratin 7/17 in Human, Bovine, Ferret (Negative), Mouse (Negative), Rabbit (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin 7/17 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| CK7, CK-7, cytokeratin 7, Cytokeratin-7, K2C7, K7keratin, 55K type II cytoskeletal, keratin 7, keratin, type II cytoskeletal 7, keratin-7, MGC129731, MGC3625, Sarcolectin, SCLkeratin, simple epithelial type I, K7, type II mesothelial keratin K7, Type-II keratin Kb7 | |
| Cytoskeletal preparation from HeLa cells (human cervical carcinoma cells) | |
| 100 μg | |
| Cell Biology, Cellular Markers | |
| 3855 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| C-46 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Bovine, Ferret (Negative), Mouse (Negative), Rabbit (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction