missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Antibody (121-3), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-78054PECY55
This item is not returnable.
View return policy
Description
DNA Monoclonal antibody specifically detects DNA in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
| DNA | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| Double stranded deoxyribonucleic acid, Double stranded DNA, dsDNA | |
| Nuclei of Burkitt's cells | |
| 0.1 mL | |
| Chromatin Research | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| 121-3 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction