missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 7, 8 Antibody (LDS65), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50459PCP
This item is not returnable.
View return policy
Description
Cytokeratin 7, 8 Monoclonal antibody specifically detects Cytokeratin 7, 8 in Human samples. It is validated for Immunohistochemistry
Specifications
| Cytokeratin 7, 8 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry | |
| LDS65 | |
| Immunohistochemistry | |
| CK7, CK-7, CK8, CK-8, Cytokeratin-7, Cytokeratin-8, keratin 8, keratin, type II cytoskeletal 7, keratin-7 | |
| Cytoskeletons of the human urothelial RT4 cell line | |
| 0.1 mL | |
| Cellular Signaling | |
| 3855 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction