missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 7, 8 Antibody (LDS68), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50460AF700
This item is not returnable.
View return policy
Description
Cytokeratin 7, 8 Monoclonal specifically detects Cytokeratin 7, 8 in Human samples. It is validated for Immunohistochemistry.
Specifications
| Cytokeratin 7, 8 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Cytoskeletons of the human urothelial RT4 cell line. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| LDS68 | |
| Immunohistochemistry | |
| Mouse | |
| Protein A purified | |
| Cellular Signaling | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction