missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 7 Antibody (rOV-TL12/30), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54499AF647
This item is not returnable.
View return policy
Description
Cytokeratin 7 Monoclonal antibody specifically detects Cytokeratin 7 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF
Specifications
| Cytokeratin 7 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| CK7, CK-7, cytokeratin 7, Cytokeratin-7, K2C7, K7keratin, 55K type II cytoskeletal, keratin 7, keratin, type II cytoskeletal 7, keratin-7, MGC129731, MGC3625, Sarcolectin, SCLkeratin, simple epithelial type I, K7, type II mesothelial keratin K7, Type-II keratin Kb7 | |
| Recombinant full-length human Cytokeratin 7 protein (Uniprot: P08729) | |
| 100 μg | |
| Cell Biology, Cellular Markers | |
| 3855 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| rOV-TL12/30 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction