Learn More
Cytokeratin 7 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-22208-100UL
Product Code. 17472523
  • € 450.00 / 100µL
Description
The Cytokeratin 7 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human Cytokeratin 7. This antibody recognizes human antigen. The Cytokeratin 7 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifications
Specifications
Cytokeratin 7
Polyclonal
Rabbit
Primary
Human
Immunofluorescence, Flow Cytometry (Intracellular Staining)
CoraLite 488
100 μL
3855