missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 8/18 Antibody (KRT8.18/1346), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54520AF647
This item is not returnable.
View return policy
Description
Cytokeratin 8/18 Monoclonal antibody specifically detects Cytokeratin 8/18 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow (Intracellular), CyTOF
Specifications
| Cytokeratin 8/18 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
| Cytoskeleton preparation from HeLa cells | |
| 100 μg | |
| Cancer, Cell Biology, Cellular Markers | |
| 3856 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining), CyTOF | |
| KRT8.18/1346 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction