missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 8/18 Antibody (KRT8/803 + KRT18/835), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47983PE
This item is not returnable.
View return policy
Description
Cytokeratin 8/18 Monoclonal specifically detects Cytokeratin 8/18 in Human samples. It is validated for Flow Cytometry.
Specifications
| Cytokeratin 8/18 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
| Mouse | |
| Protein A or G purified | |
| Cancer, Cytoskeleton Markers, Phospho Specific | |
| 3856 | |
| Human | |
| Purified |
| Flow Cytometry | |
| KRT8/803 + KRT18/835 | |
| Flow Cytometry | |
| P05787, P05783 | |
| KRT8 | |
| Recombinant full-length human KRT8 protein (KRT8/803); Recombinant full-length human KRT18 protein (KRT18/835) | |
| 0.1 mL | |
| Primary | |
| Cytokeratin 8 (CK8) belongs to the type II (or B or basic) subfamily of high molecular weight cytokeratins and exists in combination with cytokeratin 18 (CK18). This monoclonal antibody cocktail recognizes all simple epithelia including glandular epithelium, for example thyroid, female breast, gastrointestinal tract, respiratory tract, and urogenital tract including transitional epithelium. All adenocarcinomas and most squamous carcinomas are positive but keratinizing squamous carcinomas are usually negative. This antibody is useful in demonstrating the presence of Paget cells; there is very little keratin 18 in the normal epidermis so only Paget cells are stained.Immuno-histochemical staining with this monoclonal antibody is indistinguishable from that obtained with monoclonal antibody 5D3. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction