missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 8 Antibody (K8.8), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34654AF594
This item is not returnable.
View return policy
Description
Cytokeratin 8 Monoclonal antibody specifically detects Cytokeratin 8 in Human, Rat (Negative) samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
| Cytokeratin 8 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| CARD2, CK8, CK-8, CYK8, Cytokeratin-8, K2C8, K8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
| Keratin preparation from a human carcinoma (Uniprot: P05787) | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers | |
| 3856 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| K8.8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction