missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 8 Antibody (KRT8/803) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-48170
This item is not returnable.
View return policy
Description
Cytokeratin 8 Monoclonal specifically detects Cytokeratin 8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cytokeratin 8 | |
| Monoclonal | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| KRT8 | |
| Recombinant full-length human Cytokeratin 8 protein (Uniprot: P05787) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Cytokeratin 8 (CK8) belongs to the type II (or B or basic) subfamily of high molecular weight cytokeratins and exists in combination with cytokeratin 18 (CK18). CK8 is primarily found in the non-squamous epithelia and is present in majority of adenocarcinomas and ductal carcinomas. It is absent in squamous cell carcinomas. Hepatocellular carcinomas are defined by the use of antibodies that recognize only cytokeratin 8 and 18. CK8 exists on several types of normal and neoplastic epithelia, including many ductal and glandular epithelia such as colon, stomach, small intestine, trachea, and esophagus as well as in transitional epithelium. Anti-CK8 does not react with skeletal muscle or nerve cells. Epithelioid sarcoma, chordoma, and adamantinoma show strong positivity corresponding to that of simple epithelia (with antibodies against CK8, CK18 and CK19). Anti-CK8 is useful for the differentiation of lobular ('ring-like, perinuclear') from ductal ('peripheral-predominant') carcinoma of the breast. | |
| Store at 4C. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| KRT8/803 | |
| Immunohistochemistry-Paraffin | |
| CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
| Mouse | |
| 52.5 kDa | |
| 7 mL | |
| Cancer, Cytoskeleton Markers | |
| 3856 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction