missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 8 Recombinant Rabbit Monoclonal Antibody (SU0338), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen MA532118

Additional Details : Weight : 0.01000kg

Product Code. 16355544

  • € 471.00 / 100µL
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Cytokeratin 8 is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells. Cutokeratin 8 typically dimerizes with keratin 18 to form an intermediate filament in simple single-layered epithelial cells. This protein plays a role in maintaining cellular structural integrity and also functions in signal transduction and cellular differentiation. Mutations in this gene cause cryptogenic cirrhosis.
Specifications

Specifications

Cytokeratin 8
Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide, pH 7.4
P05787, P11679
KRT8
Synthetic peptide within Human Cytokeratin 8 aa 321-370
100 μL
Primary
Human, Mouse
Antibody
IgG
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot, Western Blot
SU0338
Unconjugated
KRT8
AA960620, AL022697, AU019895, CARD2, cb186, CK8, CK-8, CYK8, CYKER, cytokeratin 8, cytokeratin 8 (370 AA), cytokeratin endo A, Cytokeratin-8, cytokeratin-A, DreK8, EndoA, etID33550.23, etID43142.23, etID43142.8, K2C8, K8, KERA, keratin 8, keratin 8, type II, keratin complex 2, basic, gene 8, keratin, type 2, gene 8, Keratin, type II cytoskeletal 8, keratin, type II cytoskeletal 8-like protein, keratin-8, KO, Krt-2.8, Krt2-8, krt8, LOW QUALITY PROTEIN: keratin, type II cytoskeletal 8, sb:cb186, type II cytokeratin A (no.8), type-II keratin Kb8, wu:fa20h05, wu:fa95h10, wu:fb96c06, zfk8, zf-K8
Rabbit
Protein A
RUO
16691, 3856
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers