missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, HMW Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97591AF594
This item is not returnable.
View return policy
Description
Cytokeratin, HMW Polyclonal specifically detects Cytokeratin, HMW in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cytokeratin, HMW | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CK-2P, cytokeratin 2, Cytokeratin-2P, HUMCYT2A, K2P, K76, keratin 76, keratin, type II cytoskeletal 2 oral, Keratin-76, KRT2Bcytokeratin-2P, KRT2Pkeratin 2p, KRT76, Type-II keratin Kb9 | |
| Produced in rabbits immunized with E. coli-derived Human Cytokeratin, HMW fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 51350 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction