missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, HMW Antibody (KRTH/1576R), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-54589F
This item is not returnable.
View return policy
Description
Cytokeratin, HMW Monoclonal antibody specifically detects Cytokeratin, HMW in Human, Canine samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin, HMW | |
| Monoclonal | |
| FITC | |
| CK-2P, cytokeratin 2, Cytokeratin-2P, HUMCYT2A, K2P, K76, keratin 76, keratin, type II cytoskeletal 2 oral, Keratin-76, KRT2Bcytokeratin-2P, KRT2Pkeratin 2p, KRT76, Type-II keratin Kb9 | |
| Recombinant full-length human Cytokeratin, HMW protein (Uniprot: Q01546) | |
| 100 μg | |
| Primary | |
| Human, Canine | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| KRTH/1576R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 51350 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction