missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, HMW Antibody (rKRTH/2148), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08283AF532
This item is not returnable.
View return policy
Description
Cytokeratin, HMW Monoclonal antibody specifically detects Cytokeratin, HMW in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin, HMW | |
| Monoclonal | |
| Alexa Fluor 532 | |
| CK-2P, cytokeratin 2, Cytokeratin-2P, HUMCYT2A, K2P, K76, keratin 76, keratin, type II cytoskeletal 2 oral, Keratin-76, KRT2Bcytokeratin-2P, KRT2Pkeratin 2p, KRT76, Type-II keratin Kb9 | |
| Solubilized keratin extract from human stratum corneum | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| rKRTH/2148 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 51350 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction