missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, LMW Antibody (AE-1), Novus Biologicals™
Mouse Monoclonal Antibody
€ 229.00 - € 514.00
Specifications
| Antigen | Cytokeratin, LMW |
|---|---|
| Clone | AE-1 |
| Concentration | 0.2 mg/ml |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18423702
|
Novus Biologicals
NBP2-29411-20ug |
20 ug |
€ 229.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18164485
|
Novus Biologicals
NBP2-29411 |
0.1 mg |
€ 514.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin, LMW Monoclonal specifically detects Cytokeratin, LMW in Human, Mouse, Rat, Bovine, Canine, Chicken, Monkey, Rabbit, Reptile samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Cytokeratin, LMW | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| P04264 | |
| 374454 | |
| Human epidermal keratin (Uniprot: Q7Z794) | |
| Primary | |
| Store at 4C. |
| AE-1 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat, Bovine, Canine, Chicken, Other, Primate, Rabbit | |
| Cytokeratin LMW, cytokeratin-1B, keratin 77, keratin, type II cytoskeletal 1b, KRT77, type-II keratin Kb39 | |
| KRT77 | |
| IgG1 κ | |
| Protein A or G purified | |
| This monoclonal antibody recognizes the 56.5kDa (CK10); 50kDa (CK14); 50kDa (CK15); 48kDa (CK16); 40kDa (CK19) keratins of the acidic (Type I or LMW) subfamily. Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI, 48, 46, 45, and 40kDa. monoclonal antibody AE3 recognizes the 65-67, 64, 59, 58, 56, and 52kDa keratins of basic subfamily. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis. AE1/AE3 is a broad spectrum anti pan-keratin antibody cocktail, which differentiates epithelial tumors from non-epithelial tumors e.g. squamous vs. adenocarcinoma of the lung, liver carcinoma, breast cancer, and esophageal cancer. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title