missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, LMW Antibody (KRTL/1077), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47798H
This item is not returnable.
View return policy
Description
Cytokeratin, LMW Monoclonal antibody specifically detects Cytokeratin, LMW in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin, LMW | |
| Monoclonal | |
| HRP | |
| Cytokeratin LMW, cytokeratin-1B, keratin 77, keratin, type II cytoskeletal 1b, KRT77, type-II keratin Kb39 | |
| Recombinant human Cytokeratin, LMW protein (Uniprot: Q7Z794) | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| KRTL/1077 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 374454 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction