missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, LMW Antibody (rKRTL/6616), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08674AF488
This item is not returnable.
View return policy
Description
Cytokeratin, LMW Monoclonal antibody specifically detects Cytokeratin, LMW in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin, LMW | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Cytokeratin LMW, cytokeratin-1B, keratin 77, keratin, type II cytoskeletal 1b, KRT77, type-II keratin Kb39 | |
| Synthetic peptide corresponding to Cytokeratin, LMW residues within aa100-200 of Cytokeratin, LMW (Uniprot: Q7Z794) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| rKRTL/6616 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 374454 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction