missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin Mouse anti-Human, Rat, Clone: KRT/457, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14657.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against native human Cytokeratin.
- High specificity
- High reactivity
Specifications
| Cytokeratin | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined | |
| P02538; P04259; P13647; P19013; P48668; P05787; P13645; P13646; P05783 | |
| KRT4 | |
| Keratin-enriched preparation from cultured human epithelial cells. | |
| 100 μg | |
| Primary | |
| This antibody recognizes Type I and Type II cytokeratins, including CK4, CK5, CK6, CK8, CK10, CK13, and CK18. | |
| Store at 4°C. | |
| IgG1 |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| KRT/457 | |
| Mouse monoclonal antibody raised against native human Cytokeratin. | |
| In PBS (0.05% BSA, 0.05% sodium azide). | |
| CK4/CYK4/FLJ31692/K4 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 3851/3852/3853/3854/286887/3856/3858/3860/3875 | |
| Human, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction