missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin Mouse, Clone: 34BE12, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio MSM3-446-P1
This item is not returnable.
View return policy
Specifications
| Cytokeratin | |
| Cytoplasmic | |
| 34BE12 | |
| Unconjugated | |
| NP_056932 | |
| Mouse | |
| 50-67kDa | |
| Protein A/G Purified | |
| RUO | |
| 51350 | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Western Blot, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| KRT76 | |
| Solubilized keratin extract from human stratum corneum | |
| PC12 cells, Skin, Prostate carcinoma | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction