missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, pan Antibody (MonoPoly/4999R), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24196DL594
This item is not returnable.
View return policy
Description
Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin, pan | |
| Monoclonal | |
| DyLight 594 | |
| AEI2;CK1;EHK;EHK1;EPPK;K1;Keratin, type II cytoskeletal 1;KRT1A;NEPPK | |
| Recombinant fragments and/or synthetic peptides of human Cytokeratin, pan proteins (exact sequences are proprietary) | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
| 3848 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| MonoPoly/4999R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction