missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, pan Antibody (SPM583), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34433V
This item is not returnable.
View return policy
Description
Cytokeratin, pan Monoclonal specifically detects Cytokeratin, pan in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Guinea Pig, Goat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| pan Cytokeratin | |
| Monoclonal | |
| DyLight 405 | |
| KRT1 | |
| This Cytokeratin, pan Antibody (SPM583) was developed against keratin-enriched preparation from cultured human A431 cells. | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
| 3848 | |
| Human, Mouse, Rat, Pig, Amphibian, Bovine, Guinea Pig, Goat, Primate | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| SPM583 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This monoclonal antibody recognizes cytokeratin 4, 5, 6, 8, 10, 13, and 18. This is a broad-spectrum antibody which has been reported to differentiate epithelial tumors from non-epithelial tumors. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction