missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, pan Antibody (SPM583), Novus Biologicals™
Mouse Monoclonal Antibody
€ 274.05 - € 470.40
Specifications
| Antigen | Cytokeratin, pan |
|---|---|
| Clone | SPM583 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 ug/mL, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-4 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL, Flow (Intracellular) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18487311
|
Novus Biologicals
NBP2-34386-0.02mg |
0.02 mg |
€ 290.00 € 274.05 / 0.02mg Save € 15.95 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18477581
|
Novus Biologicals
NBP2-34386-0.1mg |
0.1 mg |
€ 498.00 € 470.40 / 0.10mg Save € 27.60 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Guinea Pig, Goat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Intracellular)Specifications
| Cytokeratin, pan | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| AEI2;CK1;EHK;EHK1;EPPK;K1;Keratin, type II cytoskeletal 1;KRT1A;NEPPK | |
| Keratin-enriched preparation from cultured human A431 | |
| Primary | |
| Store at 4C. |
| SPM583 | |
| Western Blot 1-2 ug/mL, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-4 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL, Flow (Intracellular) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig, Amphibian, Bovine, Guinea Pig, Goat, Monkey | |
| P02538, P04259, P13647, P19013, P48668, P13647 | |
| 3848 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title