missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytomegalovirus 65 kDa Late Antigen Monoclonal Antibody (B37H)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA17294
This item is not returnable.
View return policy
Description
The MA1-7294 antibody reacts with Cytomegalovirus Late Ag. in viral and infected tissue samples. MA1-7294 has been successfully used in ELISA and Immunofluorescence applications. The MA1-7294 antibody was raised against the CMV Late antigen.
Cytomegalovirus (CMV) is an opportunistic pathogen infecting lung, kidney, gut and other organs in situations where an individual is immunologically immature, such as the foetus and neonate. Infection also occurs in immunosuppressed patients e.g. transplant patients, patients undergoing chemotherapy and those with HIV infection. Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5).
Specifications
| Cytomegalovirus 65 kDa Late Antigen | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Immunocytochemistry | |
| B37H | |
| Unconjugated | |
| CMV | |
| CMV Late antigen. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction